Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF687 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501564727
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-4727 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-4727 Supplier Bethyl Laboratories, Inc Supplier No. A303279AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

The zinc finger protein 687 (ZNF687) was initially identified as a translocation partner gene with RUNX1 in patients with acute myeloid leukemia (AML). Little is known of the function of the ZNF687 protein, but it has been shown to weakly interact with the Ring1/Rnf2 RING finger protein member of the Polycomb group of proteins, suggesting it may be involved in the chromatin-modifying complexes essential for embryonic development and stem cell renewal. Other evidence suggests that ZNF687 may be part of a transcriptional network that also includes ZNF592 and ZMYMD8.

Specifications

Antigen ZNF687
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene ZNF687
Gene Accession No. Q8N1G0
Gene Alias ZNF687, Zinc finger protein 687, RP11-126K1.3
Gene Symbols ZNF687
Host Species Rabbit
Immunogen Between 100 and 150
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57592
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.