Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF689 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563117
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563117 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563117 Supplier Invitrogen™ Supplier No. PA563117
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RCFRQRWSLA VHKCSPKAPN CSPRSAIG Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 75%.

May be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF689
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF689
Gene Accession No. Q96CS4
Gene Alias 4933416E05Rik; TIPUH1; transcription-involved protein upregulated in HCC 1; Zfp689; Zinc finger protein 689; Znf689
Gene Symbols ZNF689
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF689. Recombinant protein control fragment (Product #RP-101380).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115509
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.