Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF69 Antibody (OTI3C3), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274954C
Description
ZNF69 Monoclonal specifically detects ZNF69 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
ZNF69 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ZNF69 | |
Full length recombinant protein of human ZNF69 (NP_068734) produced in E.coli. | |
0.1 mL | |
7620.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI3C3 | |
Western Blot, Immunohistochemistry | |
Cos5, hZNF3, MGC59928, zinc finger protein 69, zinc finger protein 69 (Cos5) | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction