Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF69 Monoclonal Antibody (1E3)
GREENER_CHOICE

Catalog No. PIMA524426
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524426 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524426 Supplier Invitrogen™ Supplier No. MA524426
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MPCCSHRRCR EDPGTSESQE MEEWALLDIS QRKLYKEVML ETFRNLTSVG KSWKDQNIEY EYQNPRRNFR SLIEKKVNEI K.

May be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF69
Applications ELISA
Classification Monoclonal
Clone 1E3
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene ZNF69
Gene Accession No. Q9UC07
Gene Alias Cos5; hZNF3; zinc finger protein 69; ZNF3; ZNF69
Gene Symbols ZNF69
Host Species Mouse
Immunogen ZNF69 (NP_068734.1, 1 a.a. ~ 81 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7620
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.