Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF69 Monoclonal Antibody (OTI3D11)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA526959
Description
ZNF69 Monoclonal Antibody for Western Blot, IHC (P)
May be involved in transcriptional regulation.
Specifications
ZNF69 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9UC07 | |
ZNF69 | |
Full length human recombinant protein of ZNF69 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunohistochemistry (Paraffin), Western Blot | |
OTI3D11 | |
Unconjugated | |
ZNF69 | |
Cos5; hZNF3; zinc finger protein 69; ZNF3; ZNF69 | |
Mouse | |
Affinity Chromatography | |
RUO | |
7620 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction