Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF69 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA548262
Description
ZNF69 Polyclonal Antibody for Western Blot
May be involved in transcriptional regulation.
Specifications
ZNF69 | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q9UC07 | |
ZNF69 | |
KLH conjugated synthetic peptide between 535-563 amino acids from the C-terminal region of human ZNF69. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
ZNF69 | |
Cos5; hZNF3; zinc finger protein 69; ZNF3; ZNF69 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
7620 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction