Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF690 Antibody (ZSCAN29/2610), DyLight 680, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP279881FR

Catalog No. NB011726


Only null left
Add to Cart

Description

Description

ZNF690 Monoclonal specifically detects ZNF690 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

ZNF690
Monoclonal
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
ZSCAN29
Recombinant full-length human ZNF690 protein (Uniprot: Q8IWY8)
0.1 mL
Primary
Store at 4°C in the dark.
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
ZSCAN29/2610
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
EC 2.1.1.43, EC 2.7.10.1, FLJ35867, MGC129894, MGC129895, Zfp690, zinc finger and SCAN domain containing 29, zinc finger and SCAN domain-containing protein 29, Zinc finger protein 690KOX31-like zinc finger protein, ZNF690
Mouse
Protein A or G purified
RUO
Human
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.