Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF690 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552791
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552791 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552791 Supplier Invitrogen™ Supplier No. PA552791
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RTLEQCRTKF KGLQKSYRKV KSGHPPETCP FFEEMEALMS AQVIALPSNG LEAAASHSGL VGSDAETEEP GQRGWQHEEG AEEAVAQESD SDDMDLEATP QDPNSAAPVV FRSPGGVHWG YEETKTYLAI LSETQFYEAL R Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 80%.

ZNF690 is a new candidate transcription factor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF690
Applications ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZSCAN29
Gene Accession No. Q8IWY8
Gene Alias AW125441; KOX31-like zinc finger protein; LOW QUALITY PROTEIN: zinc finger and SCAN domain-containing protein 29; RGD1559150; sim to KOX31-like Zfp; Zfp690; zinc finger and SCAN domain containing 29; zinc finger and SCAN domain containing protein 29; zinc finger and SCAN domain-containing protein 29; Zinc finger protein 690; zinc finger SCAN domains 29; ZNF690; ZSCAN29
Gene Symbols ZSCAN29
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF690. Recombinant protein control fragment (Product #RP-89276).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146050
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.