Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF691 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA539564
Description
May be involved in transcriptional regulation.
Specifications
| ZNF691 | |
| Polyclonal | |
| Unconjugated | |
| ZNF691 | |
| zinc finger protein 691, Zfp691, RP11-342M1.5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51058 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5VV52 | |
| ZNF691 | |
| A synthetic peptide derived from the internal region of human ZNF691 | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction