Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF693 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986744
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6744 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6744 Supplier Bioss Supplier No. BS5716R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene is a zinc finger transcription factor. The encoded protein may function as a tumor suppressor, and single nucleotide polymorphisms in this gene are associated with blood pressure variation. Alternative splicing results in multiple transcript variants that encode different protein isoforms.

Specifications

Antigen ZNF693
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CASZ1
Gene Accession No. Q86V15
Gene Alias 2410019P08Rik; AV096627; BC040081; BOS_15820; CAS11; castor homolog 1, zinc finger; castor zinc finger 1; Castor-related protein; Casz1; Cst; D4Ertd432e; dJ734G22.1; Kiaa3026; Putative survival-related protein; RGD1563533; sb:eu580; SRG; survival-related; wu:fk85f10; Zinc finger protein 693; Zinc finger protein castor homolog 1; ZNF693
Gene Symbols CASZ1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ZNF693/CASZ1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54897
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.