Learn More
Invitrogen™ ZNF7 Monoclonal Antibody (6F2)

Description
Peptide Sequence: DLQGAEGTEA PRTSKTDSTI RTENEQACED MDILKSESYG TVVRISPQDF PQNPGFGDVS DSEVWLDSHL GSPGLKVTGF TFQNNCLNEE TVVPKTFTK.
Specifications
Specifications
| Antigen | ZNF7 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 6F2 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | ZNF7 |
| Gene Accession No. | P17097 |
| Gene Alias | C2-H2 type zinc finger protein; HF.16; KOX4; zf30; zinc finger protein 7; zinc finger protein 7 (KOX 4, clone HF.16); zinc finger protein HF.16; Zinc finger protein KOX4; zinc finger protein zfp30; Zinc finger protein-7 (KOX4); ZNF7 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.