Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566233
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566233 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566233 Supplier Invitrogen™ Supplier No. PA566233
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QGCKELGSSGL DCQPLESQGE SAEGMSQRCE ECGKGIRATS DIALHWEINT QKISRCQECQ KKLSDCLQGK HTNNCHG Highest antigen sequence identity to the following orthologs - mouse 31%, rat 38%.

May be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF7
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF7
Gene Accession No. P17097
Gene Alias C2-H2 type zinc finger protein; HF.16; KOX4; zf30; zinc finger protein 7; zinc finger protein 7 (KOX 4, clone HF.16); zinc finger protein HF.16; Zinc finger protein KOX4; zinc finger protein zfp30; Zinc finger protein-7 (KOX4); ZNF7
Gene Symbols ZNF7
Host Species Rabbit
Immunogen Recombinant Human ZNF7. Recombinant protein control fragment (Product #RP-106914).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7553
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.