Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF70 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110608
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110608 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110608 Supplier Invitrogen™ Supplier No. PA5110608
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MEVPPATKFG ETFAFENRLE SQQGLFPGED LGDPFLQERG LEQMAVIYKE IPLGEQDEEN DDYEGNFSLC SSPVQHQSIP PGTRPQDDEL FGQTFLQKSD LSMCQIIHSE EPSPCDCAET DRGDSGPNAP HRTPQPAKPY.

The function of the ZNF70 gene has not yet been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF70
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene ZNF70
Gene Accession No. Q9UC06
Gene Alias Cos17; Zinc finger protein 70; zinc finger protein N27C7-1; ZNF70
Gene Symbols ZNF70
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-140 of human ZNF70 (NP_068735.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7621
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.