Learn More
Invitrogen™ ZNF703 Monoclonal Antibody (GT264)

Description
Positive Control: human ZNF703-transfected 293T Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | ZNF703 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | GT264 |
| Concentration | 0.21 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | Znf703 |
| Gene Accession No. | Q9H7S9 |
| Gene Alias | 1110032O19Rik; AI430822; AL022941; cb514; Csmn1; End2; ES neuronal differentiation 2; fc45d06; fk28e09; NH11; nlz; nlz1; nocA related zinc finger 1; nocA-like zinc finger protein 1; nocA-like Zn-finger protein; nocA-related zinc finger protein 1; noz1; si:dkey-158j20.1; wu:fc45d06; wu:fk28e09; Zeppo1; Zfp703; zgc:111927; Zinc finger elbow-related proline domain protein 1; zinc finger protein 703; zinc finger protein Noz1; ZNF503L; Znf703; Zpo1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.