Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Znf703 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569032
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569032 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569032 Supplier Invitrogen™ Supplier No. PA569032
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 77%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 90%.

ZNF703 belongs to the Elbow/Noc family. ZNF703 contains 1 C2H2-type zinc finger. ZNF703 may function as a transcriptional repressor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Znf703
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Zfp703
Gene Accession No. 0
Gene Alias 1110032O19Rik; AI430822; AL022941; cb514; Csmn1; End2; ES neuronal differentiation 2; fc45d06; fk28e09; NH11; nlz; nlz1; nocA related zinc finger 1; nocA-like zinc finger protein 1; nocA-like Zn-finger protein; nocA-related zinc finger protein 1; noz1; si:dkey-158j20.1; wu:fc45d06; wu:fk28e09; Zeppo1; Zfp703; zgc:111927; Zinc finger elbow-related proline domain protein 1; zinc finger protein 703; zinc finger protein Noz1; ZNF503L; Znf703; Zpo1
Gene Symbols Zfp703
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of rat Znf703.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 680717
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.