Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF703 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528640
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528640 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528640 Supplier Invitrogen™ Supplier No. PA528640
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, ZNF703-transfected 293T. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

ZNF703 belongs to the Elbow/Noc family. ZNF703 contains 1 C2H2-type zinc finger. ZNF703 may function as a transcriptional repressor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF703
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene Znf703
Gene Accession No. Q9H7S9
Gene Alias 1110032O19Rik; AI430822; AL022941; cb514; Csmn1; End2; ES neuronal differentiation 2; fc45d06; fk28e09; NH11; nlz; nlz1; nocA related zinc finger 1; nocA-like zinc finger protein 1; nocA-like Zn-finger protein; nocA-related zinc finger protein 1; noz1; si:dkey-158j20.1; wu:fc45d06; wu:fk28e09; Zeppo1; Zfp703; zgc:111927; Zinc finger elbow-related proline domain protein 1; zinc finger protein 703; zinc finger protein Noz1; ZNF503L; Znf703; Zpo1
Gene Symbols Znf703
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human ZNF703. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80139
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.