Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF705D Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | ZNF705D |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
ZNF705D Polyclonal specifically detects ZNF705D in Human samples. It is validated for Western Blot.Specifications
| ZNF705D | |
| Polyclonal | |
| Rabbit | |
| Human | |
| MGC131746, putative zinc finger protein 705C, zinc finger protein 705C, zinc finger protein 705D, ZNF705C | |
| ZNF705D | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RUO | |
| NP_001034704 | |
| 728957 | |
| Synthetic peptide directed towards the middle region of human LOC728957. Peptide sequence LGQKCYECDKSGKAFSQSSGFRGNKIIHIGEKPHACLLCGKAFSLSSDLR. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title