Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF705D Monoclonal Antibody (OTI1G1)
GREENER_CHOICE

Catalog No. PIMA527350
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527350 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527350 Supplier Invitrogen™ Supplier No. MA527350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZNF705D Monoclonal Antibody for Western Blot, IHC (P)

ZNF705D (Zinc Finger Protein 705D) is a Protein Coding gene. Among its related pathways are Gene Expression. Gene Ontology (GO) annotations related to this gene include nucleic acid binding. An important paralog of this gene is ZNF705B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF705D
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1G1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ZNF705D
Gene Accession No. P0CH99
Gene Alias putative zinc finger protein 705C; zinc finger protein 705C; zinc finger protein 705D; ZNF705C; ZNF705D
Gene Symbols ZNF705D
Host Species Mouse
Immunogen Full length human recombinant protein of ZNF705D produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 728957
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.