Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF706 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583629
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583629 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583629 Supplier Invitrogen™ Supplier No. PA583629
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KQAGQKKKQG HDQKAAAKAA LIYTCTVCRT QMPDPKTFKQ HFESKHPKTP LPPELAD Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.

ZNF706 gene ontology annotations related to this gene include negative regulation of stem cell population maintenance; negative regulation of transcription, DNA-templated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF706
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF706
Gene Accession No. Q9Y5V0
Gene Alias 3110006P09Rik; AA793007; AW536850; C77232; HSPC038; HSPC038 protein; PNAS-106; PNAS-113; Unknown (protein for IMAGE:8045247); Zfp706; zgc:92624; zgc:92624 protein; zinc finger protein 706; zinc finger protein 706 pseudogene; Znf706
Gene Symbols ZNF706
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF706. Recombinant protein control fragment (Product #RP-100130).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51123
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.