Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF708 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543609
Description
Peptide sequence: KYVKVFHKYS NAKRHKIRHT GKNPFKCKEC GKSFCMLSQL TQHEIIHTGE Sequence homology: Horse: 77%; Human: 100%.
ZNF708 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 15 C2H2-type zinc fingers and 1 KRAB domain. ZNF708 may be involved in transcriptional regulation.
Specifications
ZNF708 | |
Polyclonal | |
Unconjugated | |
ZNF708 | |
KOX8; Zinc finger protein 15; Zinc finger protein 15-like 1; zinc finger protein 15-like 1 (KOX 8); zinc finger protein 708; Zinc finger protein KOX8; ZNF15; ZNF15L1; ZNF708 | |
Rabbit | |
Affinity chromatography | |
RUO | |
7562 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P17019 | |
ZNF708 | |
Synthetic peptide directed towards the N-terminal of human ZNF708 (aa 80-129). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction