Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF71 Monoclonal Antibody (3F4)
GREENER_CHOICE

Catalog No. PIMA522424
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA522424 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA522424 Supplier Invitrogen™ Supplier No. MA522424
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MKELDPKNDI SEDKLSVVGE ATGGPTRNGA RGPGSEGVWE PGSWPERPRG DAGAEWEPLG IPQGNKLLGG SVPACHELKA FANQGCVLVP PRLDDPTEKG.

Kruppel-related zinc finger protein,endothelial zinc finger protein induced by tumor necrosis factor alpha.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF71
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 3F4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene ZNF71
Gene Accession No. Q9NQZ8
Gene Alias Endothelial zinc finger protein induced by tumor necrosis factor alpha; endothelial zinc finger protein induced by tumor necrosis factor alpha; zinc finger protein 470; EZFIT; Kruppel-related zinc finger protein; zinc finger protein 71; ZNF71
Gene Symbols ZNF71
Host Species Mouse
Immunogen ZNF71 (NP_067039.1, 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58491
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.