Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF71 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100801
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100801 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100801 Supplier Invitrogen™ Supplier No. PA5100801
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ZNF71.

Kruppel-related zinc finger protein,endothelial zinc finger protein induced by tumor necrosis factor alpha.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF71
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ZNF71
Gene Accession No. Q9NQZ8
Gene Alias Endothelial zinc finger protein induced by tumor necrosis factor alpha; endothelial zinc finger protein induced by tumor necrosis factor alpha; zinc finger protein 470; EZFIT; Kruppel-related zinc finger protein; zinc finger protein 71; ZNF71
Gene Symbols ZNF71
Host Species Rabbit
Immunogen A synthesized peptide derived from human ZNF71(Accession Q9NQZ8), corresponding to amino acid residues A21-S71.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58491
Target Species Human, Mouse, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.