Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF71 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582614
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582614 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582614 Supplier Invitrogen™ Supplier No. PA582614
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DISEDKLSVV GEATGGPTRN GARGPGSEGV WEPGSWPERP RGDAGAEWEP LGIPQGNKLL GGSVPACHEL KAFANQGCVL VPPRLDDPTE KGACPPVRRG KNFSSTSDLS KPPMPCEEKK TYDCSECGKA FSRSSSLIK Highest antigen sequence indentity to the following orthologs: Rat - 31%, Mouse - 31%.

Kruppel-related zinc finger protein,endothelial zinc finger protein induced by tumor necrosis factor alpha.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF71
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF71
Gene Accession No. Q9NQZ8
Gene Alias Endothelial zinc finger protein induced by tumor necrosis factor alpha; endothelial zinc finger protein induced by tumor necrosis factor alpha; zinc finger protein 470; EZFIT; Kruppel-related zinc finger protein; zinc finger protein 71; ZNF71
Gene Symbols ZNF71
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF71. Recombinant protein control fragment (Product #RP-91316).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58491
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.