Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF71 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | ZNF71 |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-4470
![]()
|
Proteintech Group Inc
246471AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Kruppel-related zinc finger protein,endothelial zinc finger protein induced by tumor necrosis factor alphaSpecifications
| ZNF71 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9NQZ8 | |
| 58491 | |
| ZNF71 Fusion Protein Ag18371 | |
| Primary | |
| -20°C | |
| ZNF71 |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| EZFIT, zinc finger protein 71, ZNF71 | |
| ZNF71 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title