Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF710 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556405
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556405 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556405 Supplier Invitrogen™ Supplier No. PA556405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KVKFEKVEEE EQEVYEVSVP GDDKDAGPAE APAEAASGGC DALVQSSAVK MIDLSAFSRK PRTLRHLPRT P Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 93%.

ZNF710 may be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF710
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF710
Gene Accession No. Q8N1W2
Gene Alias 5430400N05Rik; 9830165K13; AI324798; LOW QUALITY PROTEIN: zinc finger protein 710; RGD1306491; Zfp710; Zinc finger protein 710; Znf710
Gene Symbols ZNF710
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF710. Recombinant protein control fragment (Product #RP-102568).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 374655
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.