Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF711 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531815
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531815 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531815 Supplier Invitrogen™ Supplier No. PA531815
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: SK-N-SH, SK-N-SH nuclear extract, ZNF711 transfected 293T lysate. Predicted reactivity: Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a zinc finger protein of unknown function. It bears similarity to a zinc finger protein which acts as a transcriptional activator. This gene lies in a region of the X chromosome which has been associated with mental retardation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF711
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene ZNF711
Gene Accession No. Q9Y462
Gene Alias 2810409C01Rik; A230078I01Rik; CMPX1; dJ75N13.1; dJ75N13.1 (znf6-like); MRX97; Zfp711; Zinc finger protein 6; zinc finger protein 6 (CMPX1); zinc finger protein 711; ZNF4; ZNF5; ZNF6; ZNF711
Gene Symbols ZNF711
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ZNF711. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7552
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.