Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF711 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595865
Description
Immunogen sequence: SEDYLMISLD DVGEKLEHMG NTPLKIGSDG SQEDAKEDGF GSEVIKVYIF KAEAEDDVEI GGTEIVTESE YTSGHSVAGV LDQSRMQREK MVYMAVKDSS Q.
This gene encodes a zinc finger protein of unknown function. It bears similarity to a zinc finger protein which acts as a transcriptional activator. This gene lies in a region of the X chromosome which has been associated with mental retardation.
Specifications
ZNF711 | |
Polyclonal | |
Unconjugated | |
ZNF711 | |
2810409C01Rik; A230078I01Rik; CMPX1; dJ75N13.1; dJ75N13.1 (znf6-like); MRX97; Zfp711; Zinc finger protein 6; zinc finger protein 6 (CMPX1); zinc finger protein 711; ZNF4; ZNF5; ZNF6; ZNF711 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
7552 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.42 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q9Y462 | |
ZNF711 | |
A synthetic peptide corresponding to a sequence within amino acids 200-300 of human ZNF711 (NP_0013175031). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction