Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF711 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118374
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118374 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5118374 Supplier Invitrogen™ Supplier No. PA5118374
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ZNF711 Polyclonal Antibody for Western Blot, ICC/IF, IP

This gene encodes a zinc finger protein of unknown function. It bears similarity to a zinc finger protein which acts as a transcriptional activator. This gene lies in a region of the X chromosome which has been associated with mental retardation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF711
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene ZNF711
Gene Accession No. Q9Y462
Gene Alias 2810409C01Rik; A230078I01Rik; CMPX1; dJ75N13.1; dJ75N13.1 (znf6-like); MRX97; Zfp711; Zinc finger protein 6; zinc finger protein 6 (CMPX1); zinc finger protein 711; ZNF4; ZNF5; ZNF6; ZNF711
Gene Symbols ZNF711
Host Species Rabbit
Immunogen E. coli-derived Human ZNF711 fragment.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7552
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.