Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF717 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982059
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2059 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2059 Supplier Bioss Supplier No. BS12232R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen ZNF717
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ZNF717
Gene Accession No. Q9BY31
Gene Alias krueppel-like factor X17; kruppel-like zinc finger factor X17; X17; zinc finger protein 717; zinc finger protein 838; ZNF717; ZNF838
Gene Symbols ZNF717
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from Human ZNF717.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100131827
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.