Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF718 Monoclonal Antibody (OTI1D3), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA811743

Description
ZNF718 (Zinc Finger Protein 718) is a Protein Coding gene. Among its related pathways are Gene Expression. Gene Ontology (GO) annotations related to this gene include nucleic acid binding. An important paralog of this gene is ZNF595.Specifications
ZNF718 | |
Monoclonal | |
Unconjugated | |
ZNF718 | |
zinc finger protein 718 | |
Mouse | |
Affinity Chromatography | |
RUO | |
255403 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
OTI1D3 | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q3SXZ3 | |
ZNF718 | |
Full length human recombit protein of human ZNF718 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction