Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF728 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5145173
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145173 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5145173 Supplier Invitrogen™ Supplier No. PA5145173
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HELVKEPPGR TGHELWLRKL ELSLGTAIGT KVCRPASIAL NGYHSPGLWK TQDLSQTVAG RSLG Antigen sequence identity: Mouse 31%; Rat 27%.

ZNF728 is a zinc finger protein that is encoded by the ZNF728 gene in humans. It contains multiple zinc finger domains and is involved in transcriptional regulation and gene expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF728
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF728
Gene Accession No. P0DKX0
Gene Alias Zinc finger protein 728
Gene Symbols ZNF728
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Recombinant protein control fragment (Product #RP-109939).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 388523
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.