Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF732 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568705
Description
This target displays homology in the following species: Human: 100%; Mouse: 77%.
This gene encodes a kruppel-associated box-containing zinc finger protein (KRAB-ZFP). The encoded protein contains an N-terminal kruppel-associated box (KRAB) domain and sixteen C-terminal C2H2-type zinc finger domains. The KRAB-ZFPs represent the largest family of mammalian transcriptional repressors, which function through the recruitment of the nuclear co-factor KRAB-Associated Protein 1 (KAP1), to engage histone modifiers and induce heterochromatin formation.
Specifications
ZNF732 | |
Polyclonal | |
Unconjugated | |
ZNF732 | |
Zinc finger protein 732; zinc finger protein LOC654254; ZNF732 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
654254 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
B4DXR9 | |
ZNF732 | |
synthetic peptide directed towards the N-terminal region of human ZNF732. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction