Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF74 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP221047
Description
ZNF74 Polyclonal specifically detects ZNF74 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
ZNF74 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
Cos52, Zfp520, Zinc finger protein 520, zinc finger protein 74, zinc finger protein 74 (Cos52), ZNF520hZNF7 | |
Rabbit | |
72 kDa | |
0.1 mL | |
Primary | |
Human | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence | |
0.65mg/mL | |
Western Blot 1:500-1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
Q16587 | |
ZNF74 | |
Recombinant protein encompassing a sequence within the center region of human ZNF74. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
7625 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction