Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF74 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA530210
Description
Recommended positive controls: 293T, A431, Jurkat, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
May play a role in RNA metabolism.
Specifications
| ZNF74 | |
| Polyclonal | |
| Unconjugated | |
| ZNF74 | |
| 2810054M15Rik; COS52; hZNF7; KRAB8; Mszf21; mszf77; ZFP520; Zfp66; Zfp74; Zfp-74; Zinc finger protein 520; zinc finger protein 569; Zinc finger protein 74; ZNF520; Znf569; ZNF74 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7625 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.65 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q16587 | |
| ZNF74 | |
| Recombinant protein encompassing a sequence within the center region of human ZNF74. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction