Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF74 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113614
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113614 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113614 Supplier Invitrogen™ Supplier No. PA5113614
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLCAGENAST PSEPEKFPQV RRQRGAGAGE GEFVCGECGK AFRQSSSLTL For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Human: 100%.

May play a role in RNA metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF74
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF74
Gene Accession No. Q16587
Gene Alias 2810054M15Rik; COS52; hZNF7; KRAB8; Mszf21; mszf77; ZFP520; Zfp66; Zfp74; Zfp-74; Zinc finger protein 520; zinc finger protein 569; Zinc finger protein 74; ZNF520; Znf569; ZNF74
Gene Symbols ZNF74
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human ZNF74.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7625
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.