Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF740 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116334
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116334 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116334 Supplier Invitrogen™ Supplier No. PA5116334
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZNF740 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF740
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ZNF740
Gene Accession No. Q8NDX6
Gene Alias 1110034O07Rik; AW548228; OriLyt TD-element-binding protein 7; TB7; Zfp740; Zinc finger protein 740; ZNF740
Gene Symbols ZNF740
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 37-65 amino acids from the N-terminal region of human ZNF740.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283337
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.