Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF746 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554172
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554172 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554172 Supplier Invitrogen™ Supplier No. PA554172
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WNFRCKPPVG LNPRTGPEGL PYSSPDNGEA ILDPSQAPRP FNEPCKYPGR TKGFGHKPGL KKHP Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 94%.

ZNF746 is a transcription repressor that specifically binds to the 5'-TATTTT[T/G]-3' consensus sequence on promoters and represses transcription of PGC-1-alpha (PPARGC1A), thereby playing a role in regulation of neuron death.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF746
Applications ChIP Assay, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF746
Gene Accession No. Q6NUN9
Gene Alias 2810407L07Rik; AI317225; LOW QUALITY PROTEIN: zinc finger protein 746; PARIS; parkin interacting substrate; parkin-interacting substrate; parkin-interacting sustrate; Zfp746; Zinc finger protein 746; Znf746
Gene Symbols ZNF746
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF746. Recombinant protein control fragment (Product #RP-92816).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 155061
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.