Learn More
ZNF746 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | ZNF746 |
---|---|
Concentration | 0.31 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-4411
![]() ![]() |
Proteintech Group Inc
245431AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The calculated molecular weight of ZNF746 is 69 kDa, but phosphorylated ZNF746 is about 80-85 kDa.
ZNF746 may be involved in transcriptional regulation.Specifications
ZNF746 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q3U133, Q6NUN9 | |
155061, 69228 | |
ZNF746 Fusion Protein Ag21566 | |
Primary | |
-20°C | |
ZNF746 |
0.31 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
zinc finger protein 746, ZNF746 | |
Znf746 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.