Learn More
Invitrogen™ ZNF75D Polyclonal Antibody

Description
Peptide sequence: LALSEQKRIK HWKMASKLIL PESLSLLTFE DVAVYFSEEE WQLLNPLEKT Sequence homology: Dog: 86%; Horse: 86%; Human: 100%.
Specifications
Specifications
| Antigen | ZNF75D |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | ZNF75D |
| Gene Accession No. | P51815 |
| Gene Alias | D8C6; LOC100063485; Zinc finger protein 75; zinc finger protein 75 (D8C6); zinc finger protein 75D; Zinc finger protein 82; ZKSCAN24; ZN75D; ZNF75; ZNF75D; ZNF82; ZSCAN28 |
| Gene Symbols | ZNF75D |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.