Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF76 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531445
Description
Recommended positive controls: NT2D1, IMR32, U87-MG. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
May be involved in transcriptional regulation.
Specifications
ZNF76 | |
Polyclonal | |
Unconjugated | |
ZNF76 | |
2810027J07; BC025615; D6S229E; Zfp523; Zinc finger protein 523; zinc finger protein 76; zinc finger protein 76 (expressed in testis); ZNF523; Znf76 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
7629 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
P36508 | |
ZNF76 | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 264 of Human ZNF76. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction