Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF763 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543557
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543557 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543557 Supplier Invitrogen™ Supplier No. PA543557
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: KDQNIEYEYQ NPRRNFRSLI EGNVNEIKED SHCGETFTQV PDDRLNFQEK Sequence homology: Human: 100%.

ZNF763 may be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF763
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF763
Gene Accession No. Q0D2J5
Gene Alias DNA-binding protein; zinc finger protein 440 like; zinc finger protein 763; ZNF; ZNF440L; ZNF763
Gene Symbols ZNF763
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ZNF763 (aa 48-97).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284390
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.