Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF766 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590825
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590825 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590825 Supplier Invitrogen™ Supplier No. PA590825
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MMKQRTEPWT VENEMKVAKN PDRWEGIKDI NTGRSCAVRS KAGNKPITNQ LGLTFQLPLP ELEIFQGEGK IYECNQVQKF ISHSSSVSPL QRIYSGVKTH IFNKHRNDFV DFPLLSQEQK AHIRRKPYEC N; Positive Samples: Mouse kidney, Rat kidney; Cellular Location: Nucleus.

ZNF766 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 10 C2H2-type zinc fingers and 1 KRAB domain. ZNF766 may be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF766
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.14 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Alias Zinc finger protein 766; ZNF766
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 60-190 of human ZNF766 (NP_0010108511).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.