Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF767P Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144140
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144140 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144140 Supplier Invitrogen™ Supplier No. PA5144140
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZNF767P Polyclonal Antibody for IHC (P), ELISA

ZNF767P is a gene that is found in the human genome. It is an example of a gene identifier or accession number, which is used to uniquely identify specific genes in genetic databases. However, without further context or information, it is difficult to provide specific details about the function or characteristics of ZNF767P.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF767P
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene ZNF767P
Gene Accession No. Q75MW2
Gene Alias ZNF767
Gene Symbols ZNF767P
Host Species Rabbit
Immunogen Recombinant Human Protein ZNF767 protein (1-118AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79970
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.