Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF775 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5145043
Description
Immunogen sequence: GTGAGLVMKV KQEKPERLLQ TLAPQAMLVE KDKENIFQQH RGLPPRQTMG RPRALGGQEE SGSPRWAPPT EQDAGLAGRA PGSASGPLSP SLSSG Antigen sequence identity: Mouse 71%; Rat 66%.
May be involved in transcriptional regulation.
Specifications
ZNF775 | |
Polyclonal | |
Unconjugated | |
ZNF775 | |
Zinc finger protein 775; ZNF775 | |
Rabbit | |
Affinity chromatography | |
RUO | |
285971 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96BV0 | |
ZNF775 | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 9-103. Recombinant protein control fragment (Product #RP-110181). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction