Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF780A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA524037
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA524037 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA524037 Supplier Thermo Scientific Supplier No. PA524037
Add to Cart
Add to Cart

ZNF780A Polyclonal Antibody

ZNF780A Polyclonal Antibody for Western Blot, Flow

TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF780A
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ZNF780A
Gene Accession No. O75290
Gene Alias Zinc finger protein 780A; ZNF780; ZNF780A
Gene Symbols ZNF780A
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 54-83 amino acids from the N-terminal region of human ZNF780A.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284323
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.