Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF781 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | ZNF781 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
ZNF781 Polyclonal specifically detects ZNF781 in Human samples. It is validated for Western Blot.Specifications
ZNF781 | |
Polyclonal | |
Rabbit | |
Human | |
FLJ37549, MGC131783, zinc finger protein 781 | |
ZNF781 | |
IgG | |
38kDa |
Western Blot | |
Unconjugated | |
RUO | |
NP_689818 | |
163115 | |
Synthetic peptide directed towards the N terminal of human ZNF781The immunogen for this antibody is ZNF781. Peptide sequence QRNAMYLKNVAETACNFQLTQYQISHANQKPYECQICGKPFRKRAHLTQH. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title