Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF782 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552773
Description
Immunogen sequence: QSTFSVHQKV HIRAKPYEYN ECGKSCSMNS HLIWPQKSHT GEKPYECPEC GKAFSEKSRL RKHQRTHTGE KPYKCDGCDK AFSAKSGLRI HQRTHTGEKP FECHECGKSF NYKS Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 64%.
ZNF782 may be involved in transcriptional regulation.
Specifications
ZNF782 | |
Polyclonal | |
Unconjugated | |
ZNF782 | |
Zinc finger protein 782; ZNF782 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
158431 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6ZMW2 | |
ZNF782 | |
Recombinant protein corresponding to Human ZNF782. Recombinant protein control fragment (Product #RP-89434). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction