Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF784 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563766
Description
Immunogen sequence: PEAQSRSSPT PESRSQEPLD LVLVPDDCRP GTPPSDLIEI QVVKVTDTTL VPEPPEPGSF HCALCPAAFR LVSELLFHEH GHLAG Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 82%.
ZNF784 may be involved in transcriptional regulation.
Specifications
ZNF784 | |
Polyclonal | |
Unconjugated | |
ZNF784 | |
C730049F21Rik; Zfp784; Zinc finger protein 784; Znf784 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
147808 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NCA9 | |
ZNF784 | |
Recombinant protein corresponding to Human ZNF784. Recombinant protein control fragment (Product #RP-93625). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction