Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF785 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589652
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589652 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589652 Supplier Invitrogen™ Supplier No. PA589652
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MGPPLAPRPA HVPGEAGPRR TRESRPGAVS FADVAVYFSP EEWECLRPAQ RALYRDVMRE TFGHLGALGF SVPKPAFISW VEGEVEAWSP EAQDPDGESS AAFSRGQGQE AGSRDGNEEK ERLKKCPKQK EVAHEVAVKE WWPSVACPEF CNPRQSPMNP WLKDTLTRRL PHSCPDCGRN FSYPSLLASH QRVHSGERPF; Positive Samples: rat small intestine; Cellular Location: Nucleus.

The function of ZNF785 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF785
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.64 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Alias Zinc finger protein 785; zinc finger protein 785; LOW QUALITY PROTEIN: zinc finger protein 785; ZNF785
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human ZNF785 (NP_6896712).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.