Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF786 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP180405
Description
ZNF786 Polyclonal specifically detects ZNF786 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ZNF786 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
DKFZp762I137, MGC156120, zinc finger protein 786 | |
Rabbit | |
Protein A purified | |
RUO | |
136051 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1 mg/ml | |
Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
EAW80063 | |
ZNF786 | |
Synthetic peptide directed towards the N terminal of human ZNF786. Peptide sequence MAEPPRLPLTFEDVAIYFSEQEWQDLEAWQKELYKHVMRSNYETLVSLDD. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Bovine: 92%; Equine: 92%; Canine: 78%. | |
Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Rabbit | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction